Learn More
Medchemexpress LLC HY-10221 1g Medchemexpress, Vorinostat CAS:149647-78-9 Purity:>98%
SDP
Catalog No. 502028703
Encompass_Preferred
Medchemexpress, HY-10221 1g Vorinostat CAS:149647-78-9 Vorinostat (SAHA) is a potent and orally active pan-inhibitor of HDAC1, HDAC2 and HDAC3 (Class I), HDAC7 (Class II) and HDAC11 (Class IV), with ID50 values of 10 nM and 20 nM for HDAC1 and HDAC3, respectively. Vorinostat induces cell apoptosis[4]. Vorinostat is also an effective inhibitor of human papillomaviruse (HPV)-18 DNA amplification[7]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8703 Supplier Medchemexpress LLC Supplier No. HY102211G
Medchemexpress, HY-10221 1g Vorinostat CAS:149647-78-9 Vorinostat (SAHA) is a potent and orally active pan-inhibitor of HDAC1, HDAC2 and HDAC3 (Class I), HDAC7 (Class II) and HDAC11 (Class IV), with ID50 values of 10 nM and 20 nM for HDAC1 and HDAC3, respectively. Vorinostat induces cell apoptosis[4]. Vorinostat is also an effective inhibitor of human papillomaviruse (HPV)-18 DNA amplification[7]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.