Learn More
Medchemexpress LLC HY-10221 5g , Vorinostat SAHA CAS:149647-78-9 Purity:98%
SDP
Catalog No. 501492169
Encompass_Preferred
Medchemexpress, HY-10221 5g Vorinostat SAHA CAS:149647-78-9 Formula:C14H20N2O3 ID50: 10 nM (HDAC1), 20 nM (HDAC3) HDAC2, HDAC7 , HDAC11 Purity:98% Vorinostat is a potent and orally available inhibitor of HDAC1 , HDAC2 and HDAC3 (Class I) , HDAC7 (Class II) and Class IV (HDAC11) , with ID 50 values of 10 nM and 20 nM for HDAC1/3, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2169 Supplier Medchemexpress LLC Supplier No. HY102215G
Medchemexpress, HY-10221 5g Vorinostat SAHA CAS:149647-78-9 Formula:C14H20N2O3 ID50: 10 nM (HDAC1), 20 nM (HDAC3) HDAC2, HDAC7 , HDAC11 Purity:98% Vorinostat is a potent and orally available inhibitor of HDAC1 , HDAC2 and HDAC3 (Class I) , HDAC7 (Class II) and Class IV (HDAC11) , with ID 50 values of 10 nM and 20 nM for HDAC1/3, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.