Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Vorinostat (SAHA), HDAC Inhibitor, >99% Purity, 5 mg
SDP

Catalog No. 502050458
Encompass

Vorinostat (SAHA) is a histone deacetylase (HDAC) inhibitor that functions by binding to and blocking the active site of the enzyme. It inhibits both class I and class II HDACs at nanomolar concentrations, leading to the accumulation of acetylated histones and other proteins that regulate gene expression. This activity can induce cell cycle arrest, differentiation, or apoptosis in transformed cell types.

  • Inhibits class I and class II HDACs
  • Induces cell cycle arrest and apoptosis
  • Supplied as a lyophilized powder
  • Purity >99%
  • Molecular weight 264.32 g/mol
  • Soluble in DMSO at 66 mg/mL and ethanol at 2 mg/mL

Catalog No. 50-205-0458 Supplier Cell Signaling Technology Supplier No. 12520S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vorinostat (SAHA) is a histone deacetylase (HDAC) inhibitor that functions by binding to and blocking the active site of the enzyme. It inhibits both class I and class II HDACs at nanomolar concentrations, leading to the accumulation of acetylated histones and other proteins that regulate gene expression. This activity can induce cell cycle arrest, differentiation, or apoptosis in transformed cell types.

  • Inhibits class I and class II HDACs
  • Induces cell cycle arrest and apoptosis
  • Supplied as a lyophilized powder
  • Purity >99%
  • Molecular weight 264.32 g/mol
  • Soluble in DMSO at 66 mg/mL and ethanol at 2 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.