Learn More
Selleck Chemical LLC Vorinostat (SAHA) 10mM/1mL 149647-78-9 MK0683
SDP
Supplier:  Selleck Chemical LLC S104710MM/1ML
Encompass_Preferred
Vorinostat (suberoylanilide hydroxamic acid, SAHA, MK0683) is an HDAC inhibitor with IC50 of ~10 nM in a cell-free assay. Vorinostat abrogates productive HPV-18 DNA amplification. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0876790