Learn More
MSC Vornado Air Purifier, True HEPA Filtration, 3 Speed Settings, 135 sq ft
Catalog No. 5000508823
Encompass
The Vornado AC350 Air Purifier provides whole room air purification with True HEPA filtration, capturing 99.97% of airborne contaminants. It includes an activated carbon prefilter for odors and features three speed settings, making it suitable for spaces up to 135 sq ft.
  • Captures 99.97% of contaminants with HEPA filtration.
  • Removes odors and larger particles with an activated carbon prefilter.
  • Purifies air in rooms up to 135 sq ft.
  • Offers 3 speed settings and electronic push button controls.
  • Includes a filter change indicator.
  • Compact design fits easily in various spaces.
  • Operates quietly while providing powerful airflow.
  • Does not emit ozone.
  • Backed by a 5-year limited warranty.
Catalog No. 50-005-08823 Supplier MSC Supplier No. 55937742
The Vornado AC350 Air Purifier provides whole room air purification with True HEPA filtration, capturing 99.97% of airborne contaminants. It includes an activated carbon prefilter for odors and features three speed settings, making it suitable for spaces up to 135 sq ft.
  • Captures 99.97% of contaminants with HEPA filtration.
  • Removes odors and larger particles with an activated carbon prefilter.
  • Purifies air in rooms up to 135 sq ft.
  • Offers 3 speed settings and electronic push button controls.
  • Includes a filter change indicator.
  • Compact design fits easily in various spaces.
  • Operates quietly while providing powerful airflow.
  • Does not emit ozone.
  • Backed by a 5-year limited warranty.