Learn More
Research Products International Corp VORNADO Miniature Vortex Mixer, Green Cup
SDP
Catalog No. 501367907
Encompass
VORNADO MiniVortex Mixer features powerful vortexing action for tubes up to 50ml. Insta-Touch pad offers automatic motor activation when slight pressure is applied to cup head and stops when pressure is removed. Unique design prevents liquids from entering housing and prolongs motor integrity.
Catalog No. 50-136-7907 Supplier Research Products International Corp Supplier No. 400815G
VORNADO MiniVortex Mixer features powerful vortexing action for tubes up to 50ml. Insta-Touch pad offers automatic motor activation when slight pressure is applied to cup head and stops when pressure is removed. Unique design prevents liquids from entering housing and prolongs motor integrity.