Learn More
Sigma Aldrich Fine Chemicals Biosciences VornadoTM Vortexer purple1EA
Catalog No. 502901300
Encompass
Powerful is the best word to describe this mini-vortexer. With a 4 mm orbit and fixed speed of 2800 rpm the Vornado instantly vortexes even the largest samples including a nearly full 50 mL tube. Despite its powerful motor the Vornado has a footprint of less than 4X4 inches allowing it to fit on even the most crowded bench. Insta-Touch operation means that the motor is automatically activated with slight pressure on the center cup head. Operation is smooth quiet and efficient. When pressure is removed operation immediately ceases. The Vornados unique head design prevents liquids from entering the housing prolonging motor life. The unit is available with five different cup head colors.
Catalog No. 50-290-1300 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7414031EA
Powerful is the best word to describe this mini-vortexer. With a 4 mm orbit and fixed speed of 2800 rpm the Vornado instantly vortexes even the largest samples including a nearly full 50 mL tube. Despite its powerful motor the Vornado has a footprint of less than 4X4 inches allowing it to fit on even the most crowded bench. Insta-Touch operation means that the motor is automatically activated with slight pressure on the center cup head. Operation is smooth quiet and efficient. When pressure is removed operation immediately ceases. The Vornados unique head design prevents liquids from entering the housing prolonging motor life. The unit is available with five different cup head colors.