Learn More
Selleck Chemical LLC Vorolanib
SDP
Catalog No. 5000004835
Encompass_Preferred
Vorolanib (X-82 CM082) is an oral multikinase dual inhibitor of vascular endothelial growth factor receptor (VEGFR) and platelet-derived growth factor receptor (PDGFR) with antiangiogenic and antineoplastic activities
Catalog No. 50-000-04835 Supplier Selleck Chemical LLC Supplier No. S68432MG
Vorolanib (X-82 CM082) is an oral multikinase dual inhibitor of vascular endothelial growth factor receptor (VEGFR) and platelet-derived growth factor receptor (PDGFR) with antiangiogenic and antineoplastic activities