Learn More
TARGETMOL CHEMICALS INC VOROZOLE 5MG
SDP
Catalog No. 502580707
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. Vorozole (R83842) is an orally active potent and selective nonsteroidal aromatase inhibitor.Vorozole exhibits antitumor activity in vivo and may be used in the study of breast cancer. purity: 99%
Catalog No. 50-258-0707 Supplier TARGETMOL CHEMICALS INC Supplier No. T26322
Also available in 1 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. Vorozole (R83842) is an orally active potent and selective nonsteroidal aromatase inhibitor.Vorozole exhibits antitumor activity in vivo and may be used in the study of breast cancer. purity: 99%