Learn More
Cole-Parmer Argos Technologies Vortamix™ Mini Vortexer, 100-240 VAC; International
Catalog No. 501960335 Shop All Cole-Parmer Products
Encompass_Preferred
Compact vortex shaker mixes samples in centrifuge and test tubes, up to 30 mm in diameter, with little effort. The vortex motion is activated by placing a tube on the gasket cap and pressing down lightly.
Catalog No. 50-196-0335 Supplier Cole-Parmer Supplier No. 0439728
Compact vortex shaker mixes samples in centrifuge and test tubes, up to 30 mm in diameter, with little effort. The vortex motion is activated by placing a tube on the gasket cap and pressing down lightly.