Learn More
Cole-Parmer Stuart Vortex Mixer/Shaker Accessory Cup Pack
Supplier:  Cole-Parmer 0472907
Encompass_Preferred
For use with the variable-speed vortexer, pack contains: 1 x plastic cradle for standard microtitre plate 1 x circular foam insert for beakers and flasks (up to 500ml capacity) 1 x rectangular foam insert (holds 8 x 0.2ml, 8 x 0.5ml and 16 x 1.5ml microcentrifuge tubes) 1 x solid rectangular foam insert (blank for custom holes) Please note: The microtitre plate holder SA8/1 should not be used at speeds above 800rpm
Catalog No. 50-196-0946