Learn More
Sigma Aldrich Fine Chemicals Biosciences Vortex adapter for microce1EA
Catalog No. 502900870
Encompass
for use with any vortex mixer equipped with a rubber pad or test tube cup. The device holds 1.5 mL tubes in a disk mounted on a shaft. A sliding plate holds down the tube caps when in use. Provides rapid and complete mixing of many samples at once.
Catalog No. 50-290-0870 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. V07561EA
for use with any vortex mixer equipped with a rubber pad or test tube cup. The device holds 1.5 mL tubes in a disk mounted on a shaft. A sliding plate holds down the tube caps when in use. Provides rapid and complete mixing of many samples at once.