Learn More
Chemglass Life Sciences Vortex-Genie 1, 120V
SDP
Catalog No. 50143458
Encompass
Vortex-Genie 1, 120V. The Vortex Genie-1 has more violent vortexing action than any other vortex mixer on the market. It is simple to use with touch and go violent high-speed only mixing action eliminates operator variability. The Vortex Genie 1 attacks problem applications like no other vortexer can and reduces vortexing time. Has a solid metal casting and stainless steel construction. Designed not to walk like other vortexers. Available with an optional 3” platform for mixing beakers, flask or multiple tubes. Supplied with Pop-Off cap.NOTE: VORTEX-GENIE 1 MAY ONLY BE USED WITH POP-OFF CUP AND 3" PLATFORM.
Catalog No. 50-143-458 Supplier Chemglass Life Sciences Supplier No. CLS4890001
Vortex-Genie 1, 120V. The Vortex Genie-1 has more violent vortexing action than any other vortex mixer on the market. It is simple to use with touch and go violent high-speed only mixing action eliminates operator variability. The Vortex Genie 1 attacks problem applications like no other vortexer can and reduces vortexing time. Has a solid metal casting and stainless steel construction. Designed not to walk like other vortexers. Available with an optional 3” platform for mixing beakers, flask or multiple tubes. Supplied with Pop-Off cap.NOTE: VORTEX-GENIE 1 MAY ONLY BE USED WITH POP-OFF CUP AND 3" PLATFORM.