Learn More
Andwin Scientific VORTEX GENIE 2 115V
SDP
Catalog No. 50949726
Encompass_Preferred
The Vortex-Genie 2 mixes or shakes liquids in all types of container with efficient controllable speed and precision. This product features a sturdy metal housing that gives a stable platform for all kinds of mixing. Other accessories or attachments that can expand the uses of this product are sold separately.
Catalog No. 50-949-726 Supplier Andwin Scientific Supplier No. 71840
The Vortex-Genie 2 mixes or shakes liquids in all types of container with efficient controllable speed and precision. This product features a sturdy metal housing that gives a stable platform for all kinds of mixing. Other accessories or attachments that can expand the uses of this product are sold separately.