Learn More
Chemglass Life Sciences Vortex-Genie 2, 120V
SDP
Catalog No. 50143446
Encompass
Vortex-Genie 2, 120V. The Vortex Genie-2 has a variable speed control which allows slow speed shaking action up to high speed vortexing. In addition, each attachment is designed to produce different mixing actions, allowing users to choose the correct action for each application. This Genie has a broad range of attachments in various shapes, sizes, and materials which allows for mixing of almost all common tubes or containers for Hands-Free or Touch operation. Equipped with a 3-position power switch which allows continuous action or "Touch On" control. With the true "Hands-Free" operation of continuous action, operators can greatly increase lab efficiency. Designed not to walk across lab bench. Supplied with 3" platform and Pop-Off cup.
Catalog No. 50-143-446 Supplier Chemglass Life Sciences Supplier No. CLS4890002
Vortex-Genie 2, 120V. The Vortex Genie-2 has a variable speed control which allows slow speed shaking action up to high speed vortexing. In addition, each attachment is designed to produce different mixing actions, allowing users to choose the correct action for each application. This Genie has a broad range of attachments in various shapes, sizes, and materials which allows for mixing of almost all common tubes or containers for Hands-Free or Touch operation. Equipped with a 3-position power switch which allows continuous action or "Touch On" control. With the true "Hands-Free" operation of continuous action, operators can greatly increase lab efficiency. Designed not to walk across lab bench. Supplied with 3" platform and Pop-Off cup.