Learn More
USA Scientific Inc Vortex-Genie 2
Catalog No. NC9864336
Encompass
Vortex-Genie 2 variable speed vortexer with cup head and 3in platform, 120 V, 60 Hz
Catalog No. NC9864336 Supplier USA Scientific Inc Supplier No. 74045600
Vortex-Genie 2 variable speed vortexer with cup head and 3in platform, 120 V, 60 Hz