Learn More
Electron Microscopy Sciences Vortex Genie 2, Australian Plug, 230 V
SDP
Catalog No. 501899681
Encompass_Preferred
The versatile Vortex-Genie 2 mixes or shakes fluids in any type container with speed and precision. It features shaking or vortexing action with variable speed control allowing slow speed shaking action up to high speed vortexing. The broad range of attachments of various shapes, sizes, and materials allows for the mixing of common tubes or container for hands-free or touch operation.
Catalog No. 50-189-9681 Supplier Electron Microscopy Sciences Supplier No. 71840AU
The versatile Vortex-Genie 2 mixes or shakes fluids in any type container with speed and precision. It features shaking or vortexing action with variable speed control allowing slow speed shaking action up to high speed vortexing. The broad range of attachments of various shapes, sizes, and materials allows for the mixing of common tubes or container for hands-free or touch operation.