Learn More
Research Products International Corp Vortex-Genie 2, Multi-Tube Holder
SDP
Catalog No. 501977694
Encompass
Rugged and reliable, the ORIGINAL Vortex-Genie 2 is our most popular vortex mixer. Variable speed control allows high speed shaking or vortexing with tough metal housing. Supplied with 3 inch Platform (shown) and Pop-Off Cup.
Vortex-Genie 2 and 2T provide a broad range of mixing capabilities that blend fluids quickly and thoroughly with a true vortex action. Mixing results can be easily reproduced from one application to the next. Vortex-Genie 2T features automatic timing. Touch mode (1-60 sec.) and hands free (1-60 min).
Supplied with a cup head for vortexing individual tubes and a 3'' platform head designed for use with beakers or flasks. Vortex-Genie 2 and 2T feature a constant on or instatouch switch, adjustable speed control, heavy duty 9 pound cast iron base with rubber feet for stability.
Catalog No. 50-197-7694 Supplier Research Products International Corp Supplier No. 155591
Rugged and reliable, the ORIGINAL Vortex-Genie 2 is our most popular vortex mixer. Variable speed control allows high speed shaking or vortexing with tough metal housing. Supplied with 3 inch Platform (shown) and Pop-Off Cup.
Vortex-Genie 2 and 2T provide a broad range of mixing capabilities that blend fluids quickly and thoroughly with a true vortex action. Mixing results can be easily reproduced from one application to the next. Vortex-Genie 2T features automatic timing. Touch mode (1-60 sec.) and hands free (1-60 min).
Supplied with a cup head for vortexing individual tubes and a 3'' platform head designed for use with beakers or flasks. Vortex-Genie 2 and 2T feature a constant on or instatouch switch, adjustable speed control, heavy duty 9 pound cast iron base with rubber feet for stability.