Learn More
Research Products International Corp Vortex-Genie 2 Tube Holder, Vertical, 24x1.5-2.0ml
SDP
Catalog No. 501977730
Encompass
Tube Holder holds twenty-four microtubes vertically. Holder snaps on to any Vortex-Genie 2 mixer and the Vortex-Genie Pulse. No additional attachment necessary.
Catalog No. 50-197-7730 Supplier Research Products International Corp Supplier No. 155588
Tube Holder holds twenty-four microtubes vertically. Holder snaps on to any Vortex-Genie 2 mixer and the Vortex-Genie Pulse. No additional attachment necessary.