Learn More
Chemglass Life Sciences Vortex-Genie 2, Digital, 120V
SDP
Catalog No. 50143447
Encompass
Vortex-Genie 2, Digital, 120V. The Vortex-Genie 2 Digital has a variable speed control which allows slow speed shaking action up to high speed vortexing. In addition, each attachment is designed to produce different mixing actions, allowing users to choose the correct action for each application. A broad range of attachments of various shapes, sizes and materials allows for mixing of almost all common tubes or containers for hands-free or touch operation. The unit has a digital speed control and display which provides accuracy, reproducibility and repeatability. The timer function is touch on for 1-99 seconds and hands-free for 1-99 minutes or continuous. Tough metal housing combined with “just enough weight”, provides a stable platform for all types of mixing.
Catalog No. 50-143-447 Supplier Chemglass Life Sciences Supplier No. CLS48900D2
Vortex-Genie 2, Digital, 120V. The Vortex-Genie 2 Digital has a variable speed control which allows slow speed shaking action up to high speed vortexing. In addition, each attachment is designed to produce different mixing actions, allowing users to choose the correct action for each application. A broad range of attachments of various shapes, sizes and materials allows for mixing of almost all common tubes or containers for hands-free or touch operation. The unit has a digital speed control and display which provides accuracy, reproducibility and repeatability. The timer function is touch on for 1-99 seconds and hands-free for 1-99 minutes or continuous. Tough metal housing combined with “just enough weight”, provides a stable platform for all types of mixing.