Learn More
Research Products International Corp Large Tube Holder, 10 - 17 mm, Attachment for Vortex-Genie 2
SDP
Catalog No. 50212392
Encompass
The Large Ampule/Tube Attachment can be used with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse and provides vigorous end-to-end agitation combined with vortexing of up to four 10 mm - 17 mm diameter tubes or ampules.
Catalog No. 50-212-392 Supplier Research Products International Corp Supplier No. 155307
The Large Ampule/Tube Attachment can be used with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse and provides vigorous end-to-end agitation combined with vortexing of up to four 10 mm - 17 mm diameter tubes or ampules.