Learn More
Cole-Parmer Scientific Industries Vortex-Genie 2 2 Shaker, 600 to 3200 rpm, 120 VAC
Catalog No. NC9554658 Shop All Cole-Parmer Products
Encompass
Vortex-Genie 2 Shaker allows you to control speed intensity by varying pressure to the head. The variable speed shaking allows you to adjust the level of mixing from gentle shaking to vigorous vortexing. Easy-to-change platforms allow you to adapt your shaker to a variety of applications. Use the pop-off cup for single tube shaking or the 3'' platform for multiple tube or flask shaking.
Catalog No. NC9554658 Supplier Cole-Parmer Supplier No. 0472400
Vortex-Genie 2 Shaker allows you to control speed intensity by varying pressure to the head. The variable speed shaking allows you to adjust the level of mixing from gentle shaking to vigorous vortexing. Easy-to-change platforms allow you to adapt your shaker to a variety of applications. Use the pop-off cup for single tube shaking or the 3'' platform for multiple tube or flask shaking.