Learn More
Research Products International Corp Vortex-Genie 2 Tube Holder, Vertical, 6x50ml
SDP
Catalog No. 501977862
Encompass
Tube Holder holds six 50mL tubes vertically. Holder snaps on to any Vortex-Genie 2 mixer and the Vortex-Genie Pulse. No additional attachment necessary.
Catalog No. 50-197-7862 Supplier Research Products International Corp Supplier No. 155590
Tube Holder holds six 50mL tubes vertically. Holder snaps on to any Vortex-Genie 2 mixer and the Vortex-Genie Pulse. No additional attachment necessary.