Learn More
Research Products International Corp Vortex-Genie 2 Tube Holder, Horizontal, 6x50ml
SDP
Catalog No. 501977861
Encompass
Tube Holder holds six 50mL tubes horizontally. Holder snaps on to any Vortex-Genie 2 mixer and the Vortex-Genie Pulse. No additional attachment necessary.
Catalog No. 50-197-7861 Supplier Research Products International Corp Supplier No. 155587
Tube Holder holds six 50mL tubes horizontally. Holder snaps on to any Vortex-Genie 2 mixer and the Vortex-Genie Pulse. No additional attachment necessary.