Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS Vortex Genie

Catalog No. 01804431 Shop All OHAUS Products
Change view
Click to view available options
Voltage:
120 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
01-804-431 120 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-804-431 Supplier OHAUS Supplier No. 83041251
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Variable speed vortex mixer incorporates timer function for both Hands-Free and Touch Operations.

  • 1-60 sec. for Touch and 1-60 min. for Hands-Free.
  • Accepts all Vortex-Genie 2 attachments.
TRUSTED_SUSTAINABILITY

Specifications

Orbit 1 mm
Speed Range 600 to 3200 rpm
Voltage 120 V
Product Type Vortex
Model Vortex-Genie 2T
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.