Learn More
Cole-Parmer Scientific Industries Vortex-Genie 2 2 Shaker, 600 to 2700 rpm, 230 VAC
Catalog No. NC0460757 Shop All Cole-Parmer Products
Encompass
Vortex-Genie 2 Shaker allows you to control speed intensity by varying pressure to the head. The variable speed shaking allows you to adjust the level of mixing from gentle shaking to vigorous vortexing. Easy-to-change platforms allow you to adapt your shaker to a variety of applications. Use the pop-off cup for single tube shaking or the 3'' platform for multiple tube or flask shaking.
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. NC0460757 Supplier Cole-Parmer Supplier No. 0472405
Vortex-Genie 2 Shaker allows you to control speed intensity by varying pressure to the head. The variable speed shaking allows you to adjust the level of mixing from gentle shaking to vigorous vortexing. Easy-to-change platforms allow you to adapt your shaker to a variety of applications. Use the pop-off cup for single tube shaking or the 3'' platform for multiple tube or flask shaking.
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.