Learn More
Research Products International Corp Vortex-Genie, Micro-Tube Attachment, Universal
SDP
Catalog No. 501977936
Encompass
New and improved, removable Microtube Holder for use with the TurboMix Attachment on the Vortex-Genie 2 family of products, and the Vortex-Genie Pulse, as well as the Disruptor Genie. Accepts 1.5ml or 2.0ml microtubes (snap-top or screw-cap).
Catalog No. 50-197-7936 Supplier Research Products International Corp Supplier No. 155314
New and improved, removable Microtube Holder for use with the TurboMix Attachment on the Vortex-Genie 2 family of products, and the Vortex-Genie Pulse, as well as the Disruptor Genie. Accepts 1.5ml or 2.0ml microtubes (snap-top or screw-cap).