Learn More
Research Products International Corp Vortex-Genie, Micro-Tube Inserts
SDP
Catalog No. 501978179
Encompass
Two high-density foam inserts to accommodate up to 60 microtubes. This insert fits into the 6 inch (152 mm) Platform attachment. For use with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse.
Catalog No. 50-197-8179 Supplier Research Products International Corp Supplier No. 5040234
Two high-density foam inserts to accommodate up to 60 microtubes. This insert fits into the 6 inch (152 mm) Platform attachment. For use with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse.