Learn More
Research Products International Corp Vortex-Genie Pulse
SDP
Catalog No. 502229776
Encompass
Vortex-Genie Pulse takes vortex mixing to a new level. True pulsing action creates a more aggressive and random motion ideal for applications such as resuspensions, creating solutions, enzyme reactions and more. Programmable ''ON'' and ''REST'' functions are easily set and monitored using the digital controls and displays. Includes a ''Continuous'' feature that allows use as a conventional vortexer and is compatible with all Vortex-Genie accessories. Supplied with 3'' cup and platform.
Two year warranty.
Catalog No. 50-222-9776 Supplier Research Products International Corp Supplier No. 155565
Vortex-Genie Pulse takes vortex mixing to a new level. True pulsing action creates a more aggressive and random motion ideal for applications such as resuspensions, creating solutions, enzyme reactions and more. Programmable ''ON'' and ''REST'' functions are easily set and monitored using the digital controls and displays. Includes a ''Continuous'' feature that allows use as a conventional vortexer and is compatible with all Vortex-Genie accessories. Supplied with 3'' cup and platform.
Two year warranty.