Learn More
Chemglass Life Sciences Vortex Mixer I/R Kit, Low Profile, 100-240v, U.S. Plug
SDP
Catalog No. 501532377
Encompass
Vortex Mixer I/R Kit, Low Profile, 100-240v, U.S. Plug. Vortex mixer features a highly innovative and attractive design. Compact low profile mixer incorporates an infrared sensor with an adjustable mixing speed and two operation modes: Sensor or Continuous. The revolutionary IR sensor mode uses an infrared system (IR) to detect the presence of the tube and automatically starts vibrating! No press, No stress! No pressure is required to maintain vibration. The technopolymer housing and zinc base ensures optimum chemical resistance and excellent stability on the bench. Has a quiet design with its low noise motor.
Catalog No. 50-153-2377 Supplier Chemglass Life Sciences Supplier No. CLS4889V01
Vortex Mixer I/R Kit, Low Profile, 100-240v, U.S. Plug. Vortex mixer features a highly innovative and attractive design. Compact low profile mixer incorporates an infrared sensor with an adjustable mixing speed and two operation modes: Sensor or Continuous. The revolutionary IR sensor mode uses an infrared system (IR) to detect the presence of the tube and automatically starts vibrating! No press, No stress! No pressure is required to maintain vibration. The technopolymer housing and zinc base ensures optimum chemical resistance and excellent stability on the bench. Has a quiet design with its low noise motor.