Learn More
Electron Microscopy Sciences EMS BenchMixer™ Vortex Mixer, 230 V
SDP
Catalog No. 501927945
Encompass_Preferred
EMS BenchMixer vortex mixer has advanced with Q-Drive technology, the unique counter-balance system creates maximum vortexing action, while minimizing noise and excessive vibration. This results in quieter, more efficient and longer lasting operation.
Other features include "no-walk" suction feet, cold room/incubator compatibility and spill-proof electronic controls. Features powerful motor for instant vortexing and continuous or "touch" operation. Variable speed from 200 to 3200 rpm.
Catalog No. 50-192-7945 Supplier Electron Microscopy Sciences Supplier No. 8700202
EMS BenchMixer vortex mixer has advanced with Q-Drive technology, the unique counter-balance system creates maximum vortexing action, while minimizing noise and excessive vibration. This results in quieter, more efficient and longer lasting operation.
Other features include "no-walk" suction feet, cold room/incubator compatibility and spill-proof electronic controls. Features powerful motor for instant vortexing and continuous or "touch" operation. Variable speed from 200 to 3200 rpm.