Learn More
Chemglass Life Sciences Vortex Mixer I/R Kit, 115V, Complete
SDP
Catalog No. 501311360
Encompass
Vortex Mixer I/R Kit, 115V, Complete. Advanced infrared sensor vortex mixer with an adjustable mixing speed and two operation modes: Sensor or Continuous. The revolutionary IR sensor mode, an infrared system (IR) detects the presence of the test tube and the vortex mixer automatically starts vibrating! No press, No stress! No pressure is required to maintain vibration. The technopolymer structure ensures optimum chemical resistance. The ergonomic and highly innovative design along with the zinc base ensure an excellent stability on the bench.
Catalog No. 50-131-1360 Supplier Chemglass Life Sciences Supplier No. CLS488901
Vortex Mixer I/R Kit, 115V, Complete. Advanced infrared sensor vortex mixer with an adjustable mixing speed and two operation modes: Sensor or Continuous. The revolutionary IR sensor mode, an infrared system (IR) detects the presence of the test tube and the vortex mixer automatically starts vibrating! No press, No stress! No pressure is required to maintain vibration. The technopolymer structure ensures optimum chemical resistance. The ergonomic and highly innovative design along with the zinc base ensure an excellent stability on the bench.