Learn More
Electron Microscopy Sciences EMS Mortexer™ Vortex Mixer, 115 V
SDP
Catalog No. 501927941
Encompass_Preferred
The EMS Mortexer is more than your standard vortex mixer. The unique design of the Multi-Head (included) features a traditional cup head for general purpose mixing, plus holds up to eight microcentrifuge tubes. The drive system is balanced with Q-Drive technology, providing a smooth vortexing action, while minimizing noise and excessive vibration. This results in quieter, more efficient and longer lasting operation.
Increased speed, up to 3400 rpm, produces an instant vortex of even the largest sample sizes, including full 50 ml tubes. Variable speed from 200 to 3400 rpm.
Catalog No. 50-192-7941 Supplier Electron Microscopy Sciences Supplier No. 8700203
The EMS Mortexer is more than your standard vortex mixer. The unique design of the Multi-Head (included) features a traditional cup head for general purpose mixing, plus holds up to eight microcentrifuge tubes. The drive system is balanced with Q-Drive technology, providing a smooth vortexing action, while minimizing noise and excessive vibration. This results in quieter, more efficient and longer lasting operation.
Increased speed, up to 3400 rpm, produces an instant vortex of even the largest sample sizes, including full 50 ml tubes. Variable speed from 200 to 3400 rpm.