Learn More
Cole-Parmer Stuart Vortex Mixer with FREE Accessory Pack
Supplier:  Cole-Parmer 0472905
Encompass_Preferred
Vortex mixer features a robust die-cast body to prevent traveling while in use, and has an integrated port to support a lab rod for holding vessels in place during continuous operation.
Catalog No. 50-196-0407