Learn More
Electron Microscopy Sciences EMS Vornado™ Mini Vortexer 2-prong plug, 240 V, Red
SDP
Catalog No. 501927819
Encompass_Preferred
Powerful is the best word to describe this mini vortexer. With a 4 mm orbit and fixed speed of 2800 rpm, the Vornado instantly vortexes even the largest samples, including nearly full 50 ml tubes. Despite its powerful motor, the Vornado has a footprint of less than 4 x 4", allowing it to fit on even the most crowded bench.
Insta-Touch operation means that the motor is automatically activated with slight pressure on the center cup head. Operation is smooth, quiet, and efficient. When pressure is removed, operation immediately ceases.
The Vornado's unique head design prevents liquids from entering the housing, prolonging motor life.
Color: Red
Catalog No. 50-192-7819 Supplier Electron Microscopy Sciences Supplier No. 8700110
Powerful is the best word to describe this mini vortexer. With a 4 mm orbit and fixed speed of 2800 rpm, the Vornado instantly vortexes even the largest samples, including nearly full 50 ml tubes. Despite its powerful motor, the Vornado has a footprint of less than 4 x 4", allowing it to fit on even the most crowded bench.
Insta-Touch operation means that the motor is automatically activated with slight pressure on the center cup head. Operation is smooth, quiet, and efficient. When pressure is removed, operation immediately ceases.
The Vornado's unique head design prevents liquids from entering the housing, prolonging motor life.
Color: Red