Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heathrow Scientific™ Vortexer Mixer Replacement Central Pad

Catalog No. 17100466
Change view
Click to view available options
For Use With (Equipment):
Vortexer Mixer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
17-100-466 Vortexer Mixer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-100-466 Supplier Heathrow Scientific™ Supplier No. 120328
Add to Cart
Add to Cart

Replacement Central Pad for the Vortexer.

  • Vortexer Replacement Central Pad

Specifications

Product Type Replacement Central Pad
Color Black
Model 120328
For Use With (Equipment) Vortexer Mixer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.