Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heathrow Scientific™ Vortexer Mixer Replacement Multi-Head

Catalog No. 17100464
Change view
Click to view available options
Holds:
Assay Plates, Tubes, Microfuge Tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
17-100-464 Assay Plates, Tubes, Microfuge Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-100-464 Supplier Heathrow Scientific™ Supplier No. 120327
Add to Cart
Add to Cart

Replacement Multi-Head for the Vortexer.

  • Vortexer Replacement Multi-Head
  • Holds: Assay plate x 1 (to be used with retention cords), 50 mL conical tubes x 2 (horizontally mounted), 15 mL conical tubes x 2 (horizontally mounted), 5 mL tubes x 2 (horizontally mounted in the same retention space as 15 mL), 1.5/2 mL microfuge tubes x 4 (vertically mounted), 0.5 mL microfuge tubes x 6 (vertically mounted), 0.2 mL microfuge tubes (strip tubes) x 24 (vertically mounted)

Specifications

Holds Assay Plates, Tubes, Microfuge Tubes
Product Type Replacement Multi-Head
Color Black
Model 120327
For Use With (Equipment) Vortexer Mixer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.