Learn More
USA Scientific Inc Vortex-Genie 2 holder
Catalog No. 502745196
Encompass
Vortex-Genie 2 Vertical Multi-Tube Holder for 6 x 50 mL tubes
Catalog No. 50-274-5196 Supplier USA Scientific Inc Supplier No. 74049506
Vortex-Genie 2 Vertical Multi-Tube Holder for 6 x 50 mL tubes