Learn More
USA Scientific Inc Vortex-Genie 2 tube holder
Catalog No. 502745300
Encompass
Vortex-Genie 2 Vertical Multi-Tube holder for (6) 50mL tubes, (6) 15mL tubes and (12) microtubes
Catalog No. 50-274-5300 Supplier USA Scientific Inc Supplier No. 74095250
Vortex-Genie 2 Vertical Multi-Tube holder for (6) 50mL tubes, (6) 15mL tubes and (12) microtubes