Learn More
USA Scientific Inc Vortex-Genie 2T
Catalog No. 502744998
Encompass
Vortex-Genie 2T variable speed vortexer with timer, includes cup head and 3in platform, 120 V, 60 Hz
Catalog No. 50-274-4998 Supplier USA Scientific Inc Supplier No. 74082360
Vortex-Genie 2T variable speed vortexer with timer, includes cup head and 3in platform, 120 V, 60 Hz