Learn More
USA Scientific Inc Vortex-Genie Pulse
Catalog No. 502745524
Encompass
Vortex-Genie Pulse, digital display, 500-3000 rpm, 120V, 60 Hz
Catalog No. 50-274-5524 Supplier USA Scientific Inc Supplier No. 74072360
Vortex-Genie Pulse, digital display, 500-3000 rpm, 120V, 60 Hz