Learn More
Chemglass Life Sciences Vortexing Marine Style Impeller Blade for 5L & 7L Bio Reactors
SDP
Catalog No. NC1789494
Encompass
The Vortexing Marine Style Impeller Blade is used primarily for mammalian cell culture work. Stainless steel components manufactured from 316L with an electropolished surface finish of 20Ra or better.
This item is not returnable. View return policy
Catalog No. NC1789494 Supplier Chemglass Life Sciences Supplier No. CLS138205
The Vortexing Marine Style Impeller Blade is used primarily for mammalian cell culture work. Stainless steel components manufactured from 316L with an electropolished surface finish of 20Ra or better.