Learn More
Medchemexpress LLC Vortioxetine-d8 hydrobromide | 960203-27-4 | C18H15D8BrN2S | 1 MG
SDP
Catalog No. 5000376664
Encompass_Preferred
Vortioxetine-d8 hydrobromide, also known as Lu AA 21004-d8 hydrobromide, is a deuterated form of Vortioxetine. Vortioxetine acts as an antagonist of 5-HT3A and 5-HT7 receptors, and an inhibitor of the 5-hydroxytryptamine transporter (SERT). It also functions as a 5-HT1A agonist and a partial 5-HT1B agonist.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Targets 5-HT1A receptor
  • Targets 5-HT1B receptor
  • Targets 5-HT3A receptor
  • Targets 5-HT7 receptor
Catalog No. 50-003-76664 Supplier Medchemexpress LLC Supplier No. HY15414AS1MG
Vortioxetine-d8 hydrobromide, also known as Lu AA 21004-d8 hydrobromide, is a deuterated form of Vortioxetine. Vortioxetine acts as an antagonist of 5-HT3A and 5-HT7 receptors, and an inhibitor of the 5-hydroxytryptamine transporter (SERT). It also functions as a 5-HT1A agonist and a partial 5-HT1B agonist.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Targets 5-HT1A receptor
  • Targets 5-HT1B receptor
  • Targets 5-HT3A receptor
  • Targets 5-HT7 receptor