Learn More
TARGETMOL CHEMICALS INC VORTIOXETINE HYDROBROMID 100MG
SDP
Catalog No. 502577133
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 200 mg 500 mg 1000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Vortioxetine hydrobromide (Lu AA21004 hydrobromide) is a serotonin (5-HT) modulator and stimulator (SMS) inhibits 5-HT1A/1B/3A/7 receptor and SERT (IC50 15/33/3.7/19/1.6 nM). purity: 99%
Catalog No. 50-257-7133 Supplier TARGETMOL CHEMICALS INC Supplier No. T2395L100MG
Also available in 10 mg 25 mg 50 mg 200 mg 500 mg 1000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Vortioxetine hydrobromide (Lu AA21004 hydrobromide) is a serotonin (5-HT) modulator and stimulator (SMS) inhibits 5-HT1A/1B/3A/7 receptor and SERT (IC50 15/33/3.7/19/1.6 nM). purity: 99%