Learn More
Medchemexpress LLC Vortioxetine (hydrobromide) | 960203-27-4 | C18H23BrN2S | 1 G
SDP
Catalog No. 5000376661
Encompass_Preferred
Vortioxetine hydrobromide (Lu AA 21004) is a multimodal serotonergic agent that acts as an antagonist of 5-HT3A (Ki: 3.7 nM) and 5-HT7 (Ki: 19 nM) receptors. It also inhibits the 5-hydroxytryptamine transporter (SERT) with a Ki of 1.6 nM. Furthermore, it functions as a 5-HT1A agonist (Ki: 15 nM) and a partial 5-HT1B agonist (Ki: 33 nM), exhibiting potent inhibition of SERT.
  • Antagonist of 5-HT3A and 5-HT7 receptors
  • Inhibitor of 5-hydroxytryptamine transporter (SERT)
  • Acts as a 5-HT1A agonist
  • Functions as a partial 5-HT1B agonist
  • High purity of 99.87%
  • Light yellow to yellow solid appearance
Catalog No. 50-003-76661 Supplier Medchemexpress LLC Supplier No. HY15414A1G
Vortioxetine hydrobromide (Lu AA 21004) is a multimodal serotonergic agent that acts as an antagonist of 5-HT3A (Ki: 3.7 nM) and 5-HT7 (Ki: 19 nM) receptors. It also inhibits the 5-hydroxytryptamine transporter (SERT) with a Ki of 1.6 nM. Furthermore, it functions as a 5-HT1A agonist (Ki: 15 nM) and a partial 5-HT1B agonist (Ki: 33 nM), exhibiting potent inhibition of SERT.
  • Antagonist of 5-HT3A and 5-HT7 receptors
  • Inhibitor of 5-hydroxytryptamine transporter (SERT)
  • Acts as a 5-HT1A agonist
  • Functions as a partial 5-HT1B agonist
  • High purity of 99.87%
  • Light yellow to yellow solid appearance