Learn More
Medchemexpress LLC Vortioxetine hydrobromide | 960203-27-4 | C18H23BrN2S | 100 MG
SDP
Catalog No. 5000376922
Encompass_Preferred
Vortioxetine hydrobromide is a multimodal serotonergic agent acting as an antagonist of 5-HT3A and 5-HT7 receptors, and an inhibitor of 5-hydroxytryptamine transporter (SERT). It also functions as a 5-HT1A agonist and a partial 5-HT1B agonist. This compound demonstrates potent inhibition of SERT and exhibits a range of serotonergic activities.
  • Antagonist of 5-HT3A and 5-HT7 receptors
  • Inhibitor of 5-hydroxytryptamine transporter (SERT)
  • Acts as a 5-HT1A agonist and partial 5-HT1B agonist
  • Increases cell proliferation and survival
  • Stimulates maturation of immature granule cells in the hippocampus
  • Does not cause cognitive or psychomotor impairment
  • For research use only
Catalog No. 50-003-76922 Supplier Medchemexpress LLC Supplier No. HY15414A100MG
Vortioxetine hydrobromide is a multimodal serotonergic agent acting as an antagonist of 5-HT3A and 5-HT7 receptors, and an inhibitor of 5-hydroxytryptamine transporter (SERT). It also functions as a 5-HT1A agonist and a partial 5-HT1B agonist. This compound demonstrates potent inhibition of SERT and exhibits a range of serotonergic activities.
  • Antagonist of 5-HT3A and 5-HT7 receptors
  • Inhibitor of 5-hydroxytryptamine transporter (SERT)
  • Acts as a 5-HT1A agonist and partial 5-HT1B agonist
  • Increases cell proliferation and survival
  • Stimulates maturation of immature granule cells in the hippocampus
  • Does not cause cognitive or psychomotor impairment
  • For research use only