Learn More
Medchemexpress LLC Vortioxetine hydrobromide | 960203-27-4 | C18H23BrN2S | 500 MG
SDP
Catalog No. 5000376128
Encompass_Preferred
Vortioxetine hydrobromide is a multimodal serotonergic agent that acts as an antagonist for 5-HT3A and 5-HT7 receptors, an inhibitor of the 5-hydroxytryptamine transporter (SERT), a 5-HT1A agonist, and a partial 5-HT1B agonist. It is intended for research use.
  • Antagonist for 5-HT3A and 5-HT7 receptors
  • Inhibitor of 5-hydroxytryptamine transporter (SERT)
  • Agonist for 5-HT1A receptors
  • Partial agonist for 5-HT1B receptors
  • Increases cell proliferation and survival
  • Stimulates maturation of immature granule cells in hippocampus
  • Does not cause cognitive or psychomotor impairment
Catalog No. 50-003-76128 Supplier Medchemexpress LLC Supplier No. HY15414A500MG
Vortioxetine hydrobromide is a multimodal serotonergic agent that acts as an antagonist for 5-HT3A and 5-HT7 receptors, an inhibitor of the 5-hydroxytryptamine transporter (SERT), a 5-HT1A agonist, and a partial 5-HT1B agonist. It is intended for research use.
  • Antagonist for 5-HT3A and 5-HT7 receptors
  • Inhibitor of 5-hydroxytryptamine transporter (SERT)
  • Agonist for 5-HT1A receptors
  • Partial agonist for 5-HT1B receptors
  • Increases cell proliferation and survival
  • Stimulates maturation of immature granule cells in hippocampus
  • Does not cause cognitive or psychomotor impairment