Learn More
Medchemexpress LLC Vortioxetine hydrobromide | 960203-27-4 | MFCD22383961 | C18H23BrN2S | 50 MG
SDP
Catalog No. 5000376178
Encompass_Preferred
Vortioxetine hydrobromide is a multimodal serotonergic agent exhibiting antidepressant and anxiolytic potential. It functions as an antagonist of 5-HT3A and 5-HT7 receptors, an inhibitor of the 5-hydroxytryptamine transporter (SERT), a 5-HT1A agonist, and a partial 5-HT1B agonist. Research indicates its ability to increase cell proliferation and survival in the dentate gyrus of the hippocampus.
  • Antagonizes 5-HT3A and 5-HT7 receptors
  • Inhibits 5-hydroxytryptamine transporter (SERT)
  • Acts as a 5-HT1A receptor agonist
  • Functions as a partial 5-HT1B receptor agonist
  • Demonstrates antidepressant and anxiolytic properties
  • Increases cell proliferation and survival in the hippocampus
  • Does not cause cognitive or psychomotor impairment
Catalog No. 50-003-76178 Supplier Medchemexpress LLC Supplier No. HY15414A50MG
Vortioxetine hydrobromide is a multimodal serotonergic agent exhibiting antidepressant and anxiolytic potential. It functions as an antagonist of 5-HT3A and 5-HT7 receptors, an inhibitor of the 5-hydroxytryptamine transporter (SERT), a 5-HT1A agonist, and a partial 5-HT1B agonist. Research indicates its ability to increase cell proliferation and survival in the dentate gyrus of the hippocampus.
  • Antagonizes 5-HT3A and 5-HT7 receptors
  • Inhibits 5-hydroxytryptamine transporter (SERT)
  • Acts as a 5-HT1A receptor agonist
  • Functions as a partial 5-HT1B receptor agonist
  • Demonstrates antidepressant and anxiolytic properties
  • Increases cell proliferation and survival in the hippocampus
  • Does not cause cognitive or psychomotor impairment