Learn More
Apexbio Technology LLC Vortioxetine (Lu AA21004) HBr 960203-27-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000094809
Encompass_Preferred
Vortioxetine (Lu AA21004) HBr (CAS 960203-27-4) is a multimodal antidepressant that modulates the serotonergic system It acts as an antagonist at the human 5-HT3A (Ki 3 7 nM) and 5-HT7 receptors (Ki 19 nM) a partial agonist at the 5-HT1B receptor (Ki 33 nM EC50 460 nM) an agonist at 5-HT1A (Ki 15 nM) and inhibits the serotonin transporter (hSERT Ki 1 6 nM) In cellular assays it demonstrates functional antagonism at 5-HT7 and partial agonism at 5-HT1B In vivo vortioxetine modulates serotonergic activity in rodents with ED50 values of 0 4 mg/kg for rSERT and 0 11 mg/kg for 5-HT3 It is utilized in neuropharmacological studies to investigate serotonergic signaling and depression-related mechanisms
Catalog No. 50-000-94809 Supplier Apexbio Technology LLC Supplier No. A25475.1
Vortioxetine (Lu AA21004) HBr (CAS 960203-27-4) is a multimodal antidepressant that modulates the serotonergic system It acts as an antagonist at the human 5-HT3A (Ki 3 7 nM) and 5-HT7 receptors (Ki 19 nM) a partial agonist at the 5-HT1B receptor (Ki 33 nM EC50 460 nM) an agonist at 5-HT1A (Ki 15 nM) and inhibits the serotonin transporter (hSERT Ki 1 6 nM) In cellular assays it demonstrates functional antagonism at 5-HT7 and partial agonism at 5-HT1B In vivo vortioxetine modulates serotonergic activity in rodents with ED50 values of 0 4 mg/kg for rSERT and 0 11 mg/kg for 5-HT3 It is utilized in neuropharmacological studies to investigate serotonergic signaling and depression-related mechanisms