Learn More
Medchemexpress LLC Vortioxetine (Standard) | 508233-74-7 | MFCD22380814 | C18H22N2S | 100 MG
SDP
Catalog No. 5000376685
Encompass_Preferred
Vortioxetine (Lu AA 21004) is a multimodal serotonergic agent with antagonistic properties at 5-HT3A and 5-HT7 receptors, partial agonist properties at 5-HT1B receptors, and agonistic properties at 5-HT1A receptors. It also acts as a potent inhibitor of the serotonin transporter (SERT). This product is intended for research and analytical applications, and does not cause cognitive or psychomotor impairment.
  • Antagonist of 5-HT3A and 5-HT7 receptors
  • Inhibitor of serotonin transporter (SERT)
  • Agonist of 5-HT1A receptors
  • Partial agonist of 5-HT1B receptors
  • Increases cell proliferation and survival
  • Stimulates maturation of immature granule cells
  • Intended for research and analytical applications
Catalog No. 50-003-76685 Supplier Medchemexpress LLC Supplier No. HY15414100MG
Vortioxetine (Lu AA 21004) is a multimodal serotonergic agent with antagonistic properties at 5-HT3A and 5-HT7 receptors, partial agonist properties at 5-HT1B receptors, and agonistic properties at 5-HT1A receptors. It also acts as a potent inhibitor of the serotonin transporter (SERT). This product is intended for research and analytical applications, and does not cause cognitive or psychomotor impairment.
  • Antagonist of 5-HT3A and 5-HT7 receptors
  • Inhibitor of serotonin transporter (SERT)
  • Agonist of 5-HT1A receptors
  • Partial agonist of 5-HT1B receptors
  • Increases cell proliferation and survival
  • Stimulates maturation of immature granule cells
  • Intended for research and analytical applications